Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human BMP-2 Recombinant Protein
SDP

Catalog No. NB804786 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB804786 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB804786 Supplier Novus Biologicals™ Supplier No. NBP2364352UG
Add to Cart
Add to Cart

Recombinant Protein

The ED50, as determined by inducing alkaline phosphatase production using murine ATDC5 cells, is less than 200 ng/mL.

Specifications

For Use With (Application) SDS-PAGE, Bioactivity
Gene ID (Entrez) 650
Molecular Weight (g/mol) MolecularWeight-theroretical: 26 kDa
Quantity 2 μg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Endotoxin Concentration Less than 1 EU/mg of the rHuBMP-2 AS determined by LAL method.
Gene Alias BMP-2, BMP-2 A, BMP2ABone morphogenetic protein 2 A, bone morphogenetic protein 2
Gene Symbol BMP2
Reconstitution Reconstitute with 4 mM HCl to a final concentration of 0.1 mg/mL
Research Category Biologically Active Proteins, Cytokine Research, Immunology, Neuronal Cell Markers, Stem Cell Markers, Stem Cell Signaling Pathway
Purity or Quality Grade SDS-PAGE
Protein BMP-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.