Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human BMP-3 Recombinant Protein, PeproTech®

Catalog No. 12024B100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12024B100UG 100 μg
12024B10UG 10 μg
120-24B-1MG 1 mg
12024B250UG 250 μg
12024B500UG 500 μg
12024B50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 12024B100UG Supplier Gibco™ Supplier No. 12024B100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human BMP-3 is a disulfide-linked homodimeric protein that corresponds to residues 361 to 472 of the 472 amino acid BMP-3 precursor protein. The calculated molecular weight of Recombinant Human BMP-3 is 25.2 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The BMPs belong to the TGF-Beta superfamilyBMPs stimulate the production of bone matrix proteins and thereby alter stromal cell and osteoclast proliferation, a key step in bone marrow development. In addition to promoting ectopic bone and cartilage development, BMPs regulate the growth, differentiation, chemotaxis, proliferation, and apoptosis of various cell types, including mesenchymal cells, epithelial cells, hematopoietic cells, and neuronal cells. BMPs also play a role in dorsal/ventral patterning. BMP3, highly expressed in lung, ovary and small intestine, participates in cartilage and bone formation. BMP3 and BMP2 genes map to conserved regions between human and mouse.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P12645
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 651
Molecular Weight (g/mol) 25.2 kDa
Name Human BMP-3
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 9130206H07; 9530029I04Rik; BMP; Bmp3; Bmp-3; BMP3A; BMP-3 A; Bone morphogenetic protein; Bone morphogenetic protein 3; bone morphogenetic protein 3 (osteogenic); bone morphogenetic protein 3 A; bone morphogenetic protein-3; osteogenin; PBMP3
Common Name BMP-3
Gene Symbol BMP3
Biological Activity Determined by its ability to inhibit BMP-2-induced alkaline phosphatase production by ATDC-5 cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MQWIEPRNCA RRYLKVDFAD IGWSEWIISP KSFDAYYCSG ACQFPMPKSL KPSNHATIQS IVRAVGVVPG IPEPCCVPEK MSSLSILFFD ENKNVVLKVY PNMTVESCAC R
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.