Learn More
Description
- Also known as: BIP; BMP3B; BMP3b
- GDF-10 is a secreted ligand of the TGF-beta (transforming growth factor-beta) superfamily of proteins.
- Ligands of this family bind various TGF-beta receptors leading to recruitment and activation of SMAD family transcription factors that regulate gene expression.
- The preproprotein is proteolytically processed to generate each subunit of the disulfide-linked homodimer.
- This promotes neural repair after stroke.
- Additionally, GDF10 may act as a tumor suppressor and reduced expression of GDF10 is associated with oral cancer.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hGDF10 |
| Accession Number | NP_004953 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1450 bp |
| Gene ID (Entrez) | 2662 |
| Gene Alias | BIP, BMP-3b, BMP3B, bone morphogenetic protein 3B, Bone-inducing protein, GDF10, GDF-10, growth differentiation factor 10, Growth/differentiation factor 10 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
