Learn More
Description
- BMP5 bone morphogenetic protein 5 [ Homo sapiens (human) ]
- The BMP-5 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- BMPs are secreted signaling molecules that comprise a subfamily of the TGF-beta superfamily and were originally identified as regulators of cartilage and bone formation.
- There are at least 20 structurally and functionally related BMPs, most of which play roles in embryogenesis and morphogenesis of various tissues and organs.
- Biologically active BMPs are usually homodimers containing a characteristic cysteine knot structure.
- Heterodimers, BMP-2/BMP-7 and BMP-4/BMP-7 have also been suggested to exist and function in vivo. They are more potent inducers of bone formation than their respective homodimers.
- In addition, heterodimers, but not homodimers, are ventral mesoderm inducers.
- Heterodimer activity may be mediated by a different or additional receptor subtype.
- Decapentaplegic (Dpp) is one of at least five TGF-beta superfamily ligands identified in the Drosophila genome.
- Dpp, a functional ortholog of mammalian BMP-2 and BMP-4, is a morphogen and plays an essential role in Drosophila development. Dpp regulates embryonic dorsal-ventral polarity and is required for gut morphogenesis and outgrowth and patterning of imaginal disks.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hBMP5 |
| Accession Number | NP_066551 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1378 bp |
| Gene ID (Entrez) | 653 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
