Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human BMP-5 Recombinant Protein, PeproTech®

Catalog No. 12039100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 250 μg
250 μg
4 x 250 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12039100UG 100 μg
120-39-10UG 10 μg
12-039-01M 4 x 250 μg
12-039-500 2 x 250 μg
12039250UG 250 μg
120-39-2UG 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 12039100UG Supplier Gibco™ Supplier No. 12039100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

120-39-1MG will be provided as 4 x 250 μg (120-39-250UG). 120-39-500UG will be provided as 2 x 250 μg (120-39-250UG). Recombinant Human BMP-5 is a 31.2 kDa homodimeric disulfide-linked protein consisting of two identical 138 amino acid chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

TGF-β family members are key modulators of cell proliferation, differentiation, matrix synthesis, and apoptosis. As implied by their name, BMPs initiate, promote, and regulate the development, growth, and remodeling of bone and cartilage. In addition to this role, BMPs are also involved in prenatal development and postnatal growth, remodeling, and maintenance of a variety of other tissues and organs. BMP-5 is expressed in the nervous system, lungs and liver. It is a known regulator for dendritic growth in sympathetic neurons. BMP-5 is a 454 amino acid precursor protein that is cleaved to release the biologically active C-terminal mature protein.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P22003
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 653
Molecular Weight (g/mol) 31.2 kDa
Name Human BMP-5
Purification Method Purified
Quantity 100 μg
Source CHO cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias AU023399; BMP; BMP 5; Bmp5; BMP-5; bone morphogenenic protein-5; Bone morphogenetic protein; bone morphogenetic protein 5; MGC34244; se; short ear
Common Name BMP5
Gene Symbol Bmp5
Biological Activity Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 0.5-1.0 ug/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence AANKRKNQNR NKSSSHQDSS RMSSVGDYNT SEQKQACKKH ELYVSFRDLG WQDWIIAPEG YAAFYCDGEC SFPLNAHMNA TNHAIVQTLV HLMFPDHVPK PCCAPTKLNA ISVLYFDDSS NVILKKYRNM VVRSCGCH
Content And Storage -20°C
Activity Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 0.5-1.0 ug/ml.
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.