Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human BMP-7 Biotinylated, Carrier-Free Recombinant Protein

Catalog No. BT354010CF
Change view
Click to view available options
10μg; Biotin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BT354010CF 10μg; Biotin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT354010CF Supplier R&D Systems™ Supplier No. BT354010/CF
This item is not returnable. View return policy

Human Recombinant Carrier Free Protein

Specifications

Accession Number P18075
For Use With (Application) Bioactivity
Gene ID (Entrez) 655
Molecular Weight (g/mol) Theoretical M.W.: 16kDa (unlabeled)
Quantity 10μg
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt,−20 to −70°C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months,−20 to −70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU per 1μg of the protein by the LAL method.
Gene Alias BMP7, BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1, OP-1OP1, Osteogenic protein 1
Common Name BMP-7
Gene Symbol BMP7
Biological Activity Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells. Nakamura, K. et al. (1999) Exp. Cell Res. 250:351. The ED50 for this effect is typically 100-600ng/mL.
Conjugate Biotin
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Ser293-His431
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.