Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human BMPR-II ELISA Kit

Catalog No. EH50RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH50RB 96 Tests
EH50RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH50RB Supplier Invitrogen™ Supplier No. EH50RB
Add to Cart
Add to Cart

ELISA

Human BMPR-II quantitates human BMPR-II in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human BMPR-II.

BMPR2 is a member of the bone morphogenetic protein (BMP) receptor family of transmembrane serine/threonine kinases. The ligands of this receptor are BMPs that are involved in endochondral bone formation and embryogenesis. The loss of interaction and lack of phosphorylation of TCTEL1 by BMPR2 may contribute to the pathogenesis of primary pulmonary hypertension (PPH) and BMPR2 also plays an essential role in human T-cell differentiation. BMPs are involved in endochondral bone formation and embryogenesis. These proteins transduce their signals through the formation of heteromeric complexes of two different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kDa and type II receptors of about 70-80 kDa. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding. Mutations in BMPR2 have been associated with primary pulmonary hypertension, both familial and fenfluramine-associated, and with pulmonary venoocclusive disease.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 2610024H22Rik; AL117858; AW546137; BB189135; BMP type II receptor; BMP type-2 receptor; BMP-2; Bmpr2; BMPR-2; BMPR3; BMPRII; BMPR-II; BMR2; bone morphogenetic protein receptor type 2; bone morphogenetic protein receptor type II; Bone morphogenetic protein receptor type-2; bone morphogenetic protein receptor, type II (serine/threonine kinase); bone morphogenic protein receptor, type II (serine/threonine kinase); BRK-3; Gm20272; POVD1; PPH1; T-ALK; type II activin receptor-like kinase; type II receptor for bone morphogenetic protein-4
Assay Range 2.5 to 600 ng/mL
Assay Sensitivity 2.5 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 659
Gene Symbol Bmpr2
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target BMPR2
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.