Learn More
Sino Biological BPI Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 13907T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human BPI (rh BPI; Catalog#13907-H08H; P17213; Met1-Lys487). BPI specific IgG was purified by Human BPI affinity chromatography.
Catalog No. 50-255-4163
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.