Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human BRAF (NP_004324.2, 433 a.a. - 726 a.a.) T599_V600insT mutant Partial Recombinant Protein with GST-tag expressed in Baculovirus infected Sf21 cells

Catalog No. 89936851 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-936-851 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-936-851 Supplier Abnova™ Supplier No. P5756
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

This gene encodes a protein belonging to the raf/mil family of serine/threonine protein kinases. This protein plays a role in regulating the MAP kinase/ERKs signaling pathway, which affects cell division, differentiation, and secretion. Mutations in this gene are associated with cardiofaciocutaneous syndrome, a disease characterized by heart defects, mental retardation and a distinctive facial appearance. Mutations in this gene have also been associated with various cancers, including non-Hodgkin lymphoma, colorectal cancer, malignant melanoma, thyroid carcinoma, non-small cell lung carcinoma, and adenocarcinoma of lung. A pseudogene, which is located on chromosome X, has been identified for this gene. [provided by RefSeq]

Specifications

Accession Number NP_004324.2
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 673
Molecular Weight (g/mol) 60kDa
Name BRAF (T599_V600insT) (Human) Recombinant Protein
Preparation Method Baculovirus infected insect cell (Sf21) expression system
Purification Method Glutathione sepharose chromatography
Quality Control Testing SDS-PAGE Stained with Coomassie Blue
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias B-RAF1/BRAF1/FLJ95109/MGC126806/MGC138284/RAFB1
Common Name BRAF
Gene Symbol BRAF
Biological Activity The activity was determined by ELISA. The enzyme was incubated with GST-fused substrate protein, and after stopping kinase reaction by EDTA, the reaction solution was transferred into glutathione-coated plate. Phosphorylation was detected by anti-phospho antibody and HRP-labeled anti-rabbit IgG (or HRP-labeled anti-mouse IgG).
Substrate : MAP2K1 [inactive mutant]. ATP: 100 μM.
Species Baculovirus
Recombinant Recombinant
Protein Tag GST
Expression System Baculovirus infected insect cell (Sf21) expression system
Form Liquid
Purity or Quality Grade 55% by SDS-PAGE/CBB staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.