Learn More
Gibco™ Human BSEP Vesicles
Description
Human BSEP Vesicles are insect derived purified plasma membranes with inserted BSEP transport protein (bile salt export pump, ABCB11). Our Human BSEP Vesicles can be used to evaluate test compounds and drug interactions with BSEP in in vitro assays. BSEP is an ATP-binding cassette (ABC) transporter that plays a key role in bile acid excretion from the liver.
Use Human BSEP Vesicles to:
- Investigate the transporter interactions of your drug candidates
- Assess potential for transporter-mediated drug-drug interactions
- Obtain high quality results with a large signal to noise ratios
Concept Behind ABC Transporter Vesicles
ABC transporter vesicles are easy-to-use, efficient reagents for early assessment of a drug candidate's substrate and drug interaction potential. While ABC transporters typically mediate the export of substrates out of cells, transporters expressed on these inside-out vesicles import substrates into the vesicles. It is therefore possible to quantitatively evaluate transport activity for your compound by determining the amount incorporated into the vesicles.
Clear and Reliable Results
Prepared from Sf9 cells which have been engineered to over-express specific ABC transporters, these 'inside-out' vesicles provide high levels of transporter activity with low background, giving you a clear signal if your compound is a substrate or inhibitor of a specific efflux transporter.
Transporter Vesicle Details
Human BSEP Vesicles are part of a larger group of transporter vesicles known as ABC transporter vesicles. They are prepared from purified plasma membranes isolated from an insect cell system (Sf9 cells transfected with baculovirus) expressing BSEP. Evaluation of the inhibitory activity of newly developed compounds towards BSEP-mediated transport of bile acids (e.g. taurocholate) may give an indication of potential hepatotoxicity of the compounds.
For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
Specifications
Specifications
| Quantity | 0.5 mL |
| Purity | Research Grade |
| Host Species | Human |
| Product Type | Human Vesicle |
| Content And Storage | Store at -80°C. |
| Format | Vial |
| Shipping Condition | Dry Ice |
| Physical Form | Frozen |
Frequently Asked Questions (FAQs)
The quantity supplied is 2.5 mg total protein/500 µL per vial. The protein concentration is 5 mg/mL. When our recommended protocol is followed, 50 vesicular transport assays or 125 ATPase assays can be performed with one vial of ABC Transporter Vesicles product.
We recommend storing them at -80 degrees C.
Prepared from Sf9 cells which have been engineered to over-express specific ABC transporters, ATP-binding cassette (ABC) Transporter Vesicles are inside-out vesicles designed for use in vesicular transport assays. They allow the investigation of possible interactions between specific ABC transporters and test compounds. ABC Transporter Vesicles provide high levels of transporter activity with low background, giving a clear signal if the test compound is a substrate or inhibitor of a specific efflux transporter.
While ABC transporters typically mediate the efflux of substrates from cells, transporters expressed on these inside-out vesicles import substrates into the vesicles. It is therefore possible to quantitatively evaluate transport activity for the test compound by determining the amount incorporated into the vesicles.
Note: One vial of vesicle product can perform 50 vesicular transport assays.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.