Learn More
ACROBiosystems Human BTN3A1 25ug
Supplier:  ACROBiosystems BT1H82E425UG
Encompass_Preferred
Biotinylated Human BTN3A1 Protein HisAvitag (BT1-H82E4) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Gly 254 (Accession # O00481-1).
Catalog No. 50-253-5457
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.