Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human BTN3A2 Full-length ORF (NP_008978.2, 1 a.a. - 334 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89937003 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-937-003 10 μg
89-937-004 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-937-003 Supplier Abnova™ Supplier No. H00011118P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the immunoglobulin superfamily, containing two Ig domains with similarity to Ig variable and Ig constant domains. The gene resides in the juxta-telomeric region of the major histocompatability class 1 locus on chromosome 6 in the seven member BTN cluster, which includes butyrophilin, and three members each of the BTN2 and BTN3 subfamilies. Alternatively spliced transcript variants have been described but their full-length nature has yet to be determined. [provided by RefSeq]

Sequence: MKMASSLAFLLLNFHVSLLLVQLLTPCSAQFSVLGPSGPILAMVGEDADLPCHLFPTMSAETMELKWVSSSLRQVVNVYADGKEVEDRQSAPYRGRTSILRDGITAGKAALRIHNVTASDSGKYLCYFQDGDFYEKALVELKVAALGSNLHVEVKGYEDGGIHLECRSTGWYPQPQIQWSNAKGENIPAVEAPVVADGVGLYEVAASVIMRGGSGEGVSCIIRNSLLGLEKTASISIADPFFRSAQPWIAALAGTLPILLLLLAGASYFLWRQQKEITALSSEIESEQEMKEMGYAATEREISLRESLQEELKRKKIQYLTRGEESSSDTNKSA

Specifications

Accession Number NP_008978.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 11118
Molecular Weight (g/mol) 62.8kDa
Name BTN3A2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BT3.2/BT3.3/BTF4
Common Name BTN3A2
Gene Symbol BTN3A2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.