Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human BTN3A3 Fc Chimera Recombinant Protein

Catalog No. PIRP87962
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP87962 100 μg
PIRP87963 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIRP87962 Supplier Invitrogen™ Supplier No. RP87962
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. In general, recombinant proteins are provided as lyophilized powder.

Plays a role in T-cell responses in the adaptive immune response.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O00478
For Use With (Application) Control, Neutralization
Format Lyophilized
Formulation PBS with 0.01% mannitol, 5-8% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 10384
Molecular Weight (g/mol) 53 kDa
Name Human BTN3A3 Fc Chimera
pH Range 7.4
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias BTF3; BTN3.3; BTN3A3; butyrophilin 3; butyrophilin subfamily 3 member A3; butyrophilin subfamily 3 member A3 secreted isoform; butyrophilin, subfamily 3, member A3
Common Name BTN3A3
Gene Symbol BTN3A3
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence Human BNT3A3, amino acids Met1-Trp248 (Accession # O00478-1) with a C-terminal mouse IgG1 Fc region
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.