Learn More
Abcam Human c-Myc Peptide, Synthetic, Lyophilized, 1 mg
Description
- Target Protein: c-Myc
- Species Reactivity: Human
- Application: BL
- Form: Lyophilized
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | c-Myc (Human) |
| Substrate Properties | Peptide-based Substrate |
| Reactivity | Human |
| Shipping Condition | Blue Ice |
| Form | Lyophilized |
| Label or Dye | Unconjugated |
| Content And Storage | Store at -20°C. |
| Cross Reactivity | Human |
| Quantity | 1 mg |
| For Use With (Application) | Blocking, Neutralization |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.