Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human C-Peptide Quantikine ELISA Kit
SDP

Catalog No. DICP00 Shop All R&D Systems Products
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
DICP00 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DICP00 Supplier R&D Systems™ Supplier No. DICP00
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

a 2.5 hour solid-phase ELISA designed to measure human Insulin C-Peptide in cell culture supernates, serum, plasma, and urine.

The Human C-Peptide Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human C-Peptide in cell culture supernates (100 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), urine (10 ul).

Specifications

Assay Range 28.90 - 1850 pmol/L
Assay Sensitivity 2.88 pmol/L
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and synthetic Human Insulin C-Peptide
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 3630
Sample Volume Cell Culture Supernates (100uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Urine (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym C-P, IDDM2, ILPR, insulin, Insulin C-Peptide, IRDN, MODY10, proinsulin, Proinsulin C-Peptide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.