Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human C-Reactive Protein/CRP Quantikine ELISA Kit

Catalog No. DCRP00
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
DCRP00 1 Plate 1 x 96-well Plate
SCRP00 6 Plates 6 x 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DCRP00 Supplier R&D Systems™ Supplier No. DCRP00
This item is not returnable. View return policy

Human C-Reactive Protein/CRP Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human C-Reactive Protein/CRP Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human C-Reactive Protein/CRP in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).

Specifications

Assay Range 0.78 - 50ng/mL
Assay Sensitivity 0.022ng/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Natural and Recombinant Human CRP
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 1401
Quantity 1 Plate
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.