Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human C-Reactive Protein/CRP QuicKit Control Group 265

Catalog No. QC265
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QC265 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. QC265 Supplier R&D Systems™ Supplier No. QC265
Add to Cart
Add to Cart
This item is not returnable. View return policy

For use with catalog number QK1707

Specifications

Target Species Human
For Use With (Application) Control
Gene ID (Entrez) 1401
Quantity 1 Ea.
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895
Target C-Reactive Protein/CRP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.