Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human C10orf97 Full-length ORF (NP_079224.1, 1 a.a. - 445 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89937082 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-937-082 10 μg
89-937-083 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-937-082 Supplier Abnova™ Supplier No. H00080013P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

C10ORF97 is a member of the caspase-associated recruitment domain (CARD) family of proteins that play a role in apoptosis (Liu et al., 2002 [PubMed 12054670]).[supplied by OMIM]

Sequence: MSELTKELMELVWGTKSSPGLSDTIFCRWTQGFVFSESEGSALEQFEGGPCAVIAPVQAFLLKKLLFSSEKSSWRDCSEEEQKELLCHTLCDILESACCDHSGSYCLVSWLRGKTTEETASISGSPAESSCQVEHSSALAVEELGFERFHALIQKRSFRSLPELKDAVLDQYSMWGNKFGVLLFLYSVLLTKGIENIKNEIEDASEPLIDPVYGHGSQSLINLLLTGHAVSNVWDGDRECSGMKLLGIHEQAAVGFLTLMEALRYCKVGSYLKSPKFPIWIVGSETHLTVFFAKDMALVAPEAPSEQARRVFQTYDPEDNGFIPDSLLEDVMKALDLVSDPEYINLMKNKLDPEGLGIILLGPFLQEFFPDQGSSGPESFTVYHYNGLKQSNYNEKVMYVEGTAVVMGFEDPMLQTDDTPIKRCLQTKWPYIELLWTTDRSPSLN

Specifications

Accession Number NP_079224.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 80013
Molecular Weight (g/mol) 76.1kDa
Name C10orf97 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MSELTKELMELVWGTKSSPGLSDTIFCRWTQGFVFSESEGSALEQFEGGPCAVIAPVQAFLLKKLLFSSEKSSWRDCSEEEQKELLCHTLCDILESACCDHSGSYCLVSWLRGKTTEETASISGSPAESSCQVEHSSALAVEELGFERFHALIQKRSFRSLPELKDAVLDQYSMWGNKFGVLLFLYSVLLTKGIENIKNEIEDASEPLIDPVYGHGSQSLINLLLTGHAVSNVWDGDRECSGMKLLGIHEQAAVGFLTLMEALRYCKVGSYLKSPKFPIWIVGSETHLTVFFAKDMALVAPEAPSEQARRVFQTYDPEDNGFIPDSLLEDVMKALDLVSDPEYINLMKNKLDPEGLGIILLGPFLQEFFPDQGSSGPESFTVYHYNGLKQSNYNEKVMYVEGTAVVMGFEDPMLQTDDTPIKRCLQTKWPYIELLWTTDRSPSLN
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CARP/DERP5/FLJ13397/MST126/MSTP126/RP11-394I23.1/my042
Common Name C10orf97
Gene Symbol C10orf97
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.