Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human C14orf166 (NM_016039) Full-length Recombinant Protein expressed in yeast

Catalog No. 89937177 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-937-177 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-937-177 Supplier Abnova™ Supplier No. P3734
Add to Cart
Add to Cart

Used for SDS-PAGE

Sequence: MFRRKLTALDYHNPAGFNCKDETEFRNFIVWLEDQKIRHYKIEDRGNLRNIHSSDWPKFFEKYLRDVNCPFKIQDRQEAIDWLLGLAVRLEYGDNAEKYKDLVPDNSKTADNATKNAEPLINLDVNNPDFKAGVMALANLLQIQRHDDYLVMLKAIRILVQERLTQDAVAKANQTKEGLPVALDKHILGFDTGDAVLNEAAQILRLLHIEELRELQTKINEAIVAVQAIIADPKTDHRLGKVGR

Specifications

Accession Number NM_016039
For Use With (Application) SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 51637
Molecular Weight (g/mol) 28kDa
Name C14orf166 (Human) Recombinant Protein
Preparation Method Yeast expression system
Purification Method Affinity Purification
Quantity 100 μg
Immunogen MFRRKLTALDYHNPAGFNCKDETEFRNFIVWLEDQKIRHYKIEDRGNLRNIHSSDWPKFFEKYLRDVNCPFKIQDRQEAIDWLLGLAVRLEYGDNAEKYKDLVPDNSKTADNATKNAEPLINLDVNNPDFKAGVMALANLLQIQRHDDYLVMLKAIRILVQERLTQDAVAKANQTKEGLPVALDKHILGFDTGDAVLNEAAQILRLLHIEELRELQTKINEAIVAVQAIIADPKTDHRLGKVGR
Storage Requirements Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CGI-99/CGI99/CLE/CLE7/LCRP369/RLLM1
Common Name C14orf166
Gene Symbol C14orf166
Species Yeast
Recombinant Recombinant
Protein Tag None
Expression System Yeast expression system
Form Liquid
Purity or Quality Grade 0.9
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.