Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human C19orf2 Full-length ORF (NP_604431.1, 1 a.a. - 495 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89937266 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-937-266 25 μg
89-937-265 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-937-266 Supplier Abnova™ Supplier No. H00008725P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene binds to RNA polymerase II subunit 5 (RPB5) and negatively modulates transcription through its binding to RPB5. The encoded protein seems to have inhibitory effects on various types of activated transcription, but it requires the RPB5-binding region. This protein acts as a corepressor. It is suggested that it may require signaling processes for its function or that it negatively modulates genes in the chromatin structure. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]

Sequence: MWRGCARSRKRKKVDNDYNALRERLSTLPDKLSYNIMVPFGPFAFMPGKLVHTNEVTVLLGDNWFAKCSAKQAVGLVEHRKEHVRKTIDDLKKVMKNFESRVEFTEDLQKMSDAAGDIVDIREEIKCDFEFKAKHRIAHKPHSKPKTSDIFEADIANDVKSKDLLADKELWARLEELERQEELLGELDSKPDTVIANGEDTTSSEEEKEDRNTNVNAMHQVTDSHTPCHKDVASSEPFSGQVNSQLNCSVNGSSSYHSDDDDDDDDDDDDDNIDDDDGDNDHEALGVGDNSIPTIYFSHTVEPKRVRINTGKNTTLKFSEKKEEAKRKRKNSTGSGHSAQELPTIRTPADIYRAFVDVVNGEYVPRKSILKSRSRENSVCSDTSESSAAEFDDRRGVLRSISCEEATCSDTSESILEEEPQENQKKLLPLSVTPEAFSGTVIEKEFVSPSLTPPPAIAHPALPTIPERKEVLLEASEETGKRVSKFKAARLQQKD

Specifications

Accession Number NP_604431.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8725
Molecular Weight (g/mol) 82kDa
Name C19orf2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MWRGCARSRKRKKVDNDYNALRERLSTLPDKLSYNIMVPFGPFAFMPGKLVHTNEVTVLLGDNWFAKCSAKQAVGLVEHRKEHVRKTIDDLKKVMKNFESRVEFTEDLQKMSDAAGDIVDIREEIKCDFEFKAKHRIAHKPHSKPKTSDIFEADIANDVKSKDLLADKELWARLEELERQEELLGELDSKPDTVIANGEDTTSSEEEKEDRNTNVNAMHQVTDSHTPCHKDVASSEPFSGQVNSQLNCSVNGSSSYHSDDDDDDDDDDDDDNIDDDDGDNDHEALGVGDNSIPTIYFSHTVEPKRVRINTGKNTTLKFSEKKEEAKRKRKNSTGSGHSAQELPTIRTPADIYRAFVDVVNGEYVPRKSILKSRSRENSVCSDTSESSAAEFDDRRGVLRSISCEEATCSDTSESILEEEPQENQKKLLPLSVTPEAFSGTVIEKEFVSPSLTPPPAIAHPALPTIPERKEVLLEASEETGKRVSKFKAARLQQKD
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ10575/NNX3/RMP/URI
Common Name C19orf2
Gene Symbol C19orf2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.