Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human C4B Partial ORF (NP_000343, 774 a.a. - 873 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89937529 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-937-529 10 ug
89-937-530 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-937-529 Supplier Abnova™ Supplier No. H00000721Q02S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes the basic form of complement factor 4, part of the classical activation pathway. The protein is expressed as a single chain precursor which is proteolytically cleaved into a trimer of alpha, beta, and gamma chains prior to secretion. The trimer provides a surface for interaction between the antigen-antibody complex and other complement components. The alpha chain may be cleaved to release C4 anaphylatoxin, a mediator of local inflammation. Deficiency of this protein is associated with systemic lupus erythematosus. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. Varying haplotypes of this gene cluster exist, such that individuals may have 1, 2, or 3 copies of this gene. In addition, this gene exists as a long form and a short form due to the presence or absence of a 6.4 kb endogenous HERV-K retrovirus in intron 9. [provided by RefSeq]

Sequence: VRSFFPENWLWRVETVDRFQILTLWLPDSLTTWEIHGLSLSKTKGLCVATPVQLRVFREFHLHLRLPMSVRRFEQLELRPVLYNYLDKNLTVSVHVSPVE

Specifications

Accession Number NP_000343
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 721
Molecular Weight (g/mol) 36.74kDa
Name C4B (Human) Recombinant Protein (Q02)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen VRSFFPENWLWRVETVDRFQILTLWLPDSLTTWEIHGLSLSKTKGLCVATPVQLRVFREFHLHLRLPMSVRRFEQLELRPVLYNYLDKNLTVSVHVSPVE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias C4A/C4A13/C4A91/C4B1/C4B12/C4B2/C4B3/C4B5/C4F/CH/CO4/CPAMD3/FLJ60561/MGC164979
Common Name C4B
Gene Symbol C4B
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.