Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human C6orf25 Full-length ORF (NP_079536.2, 1 a.a. - 237 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89937583 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-937-583 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-937-583 Supplier Abnova™ Supplier No. H00080739P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is a member of the immunoglobulin (Ig) superfamily and is located in the major histocompatibility complex (MHC) class III region. The protein encoded by this gene is a glycosylated, plasma membrane-bound cell surface receptor, but soluble isoforms encoded by some transcript variants have been found in the endoplasmic reticulum and Golgi before being secreted. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MAVFLQLLPLLLSRAQGNPGASLDGRPGDRVNLSCGGVSHPIRWVWAPSFPACKGLSKGRRPILWASSSGTPTVPPLQPFVGRLRSLDSGIRRLELLLSAGDSGTFFCKGRHEDESRTVLHVLGDRTYCKAPGPTHGSVYPQLLIPLLGAGLVLGLGALGLVWWLHRRLPPQPIRPLPRFALSPPHSSTCENRAPEASKGGRAQDSRGPGPGTEPALCGSGPSSPQQAPPAVHSGPC

Specifications

Accession Number NP_079536.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 80739
Molecular Weight (g/mol) 51.4kDa
Name C6orf25 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MAVFLQLLPLLLSRAQGNPGASLDGRPGDRVNLSCGGVSHPIRWVWAPSFPACKGLSKGRRPILWASSSGTPTVPPLQPFVGRLRSLDSGIRRLELLLSAGDSGTFFCKGRHEDESRTVLHVLGDRTYCKAPGPTHGSVYPQLLIPLLGAGLVLGLGALGLVWWLHRRLPPQPIRPLPRFALSPPHSSTCENRAPEASKGGRAQDSRGPGPGTEPALCGSGPSSPQQAPPAVHSGPC
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias G6b/MGC142279/MGC142281/NG31
Common Name C6orf25
Gene Symbol C6orf25
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.