Learn More
Description
Specifications
Specifications
| Assay Range | 0.50 - 32 U/mL |
| Assay Sensitivity | 0.099 U/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum, Urine |
| Specificity | Natural and Recombinant Human CA125 |
| Sufficient For | 1 x 96-well Plate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 94025 |
| Sample Volume | Cell Culture Supernates (100uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Saliva (10uL),Urine (100uL),Human Milk (10uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
