Learn More
Sino Biological Human Carbonic Anhydrase XIV / CA14 Protein (His Tag), Biotinylated 20ug
Supplier:  Sino Biological 10458H08HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CA14 (NP_036245.1) (Met1-Met290) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5292
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.