Learn More
Sino Biological Human CA5A PAb 100UL 10500-RP02-100
Supplier:  Sino Biological 10500RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Carbonic Anhydrase VA / CA5A (rh Carbonic Anhydrase VA / CA5A; Catalog#10500-H08E; NP_001730.1; Ala 40-Ser 305). Carbonic Anhydrase VA / CA5A specific IgG was purified by Human Carbonic Anhydrase VA / CA5A affinity chromatography.
Catalog No. 50-240-8977
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.