Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Cadherin 13 ELISA Kit

Catalog No. EH60RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH60RB 96 Tests
EH60RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH60RB Supplier Invitrogen™ Supplier No. EH60RB
Add to Cart
Add to Cart

ELISA

Human Cadherin 13 quantitates human Cadherin 13 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cadherin 13.

The protein encoded by this gene has a C-terminal domain with tRNA 3 processing endoribonuclease activity, which catalyzes the removal of the 3' trailer from precursor tRNAs. The protein also interacts with activated Smad family member 2 (Smad2) and its nuclear partner forkhead box H1 (also known as FAST-1), and reduced expression can suppress transforming growth factor-beta induced growth arrest. Mutations in this gene result in an increased risk of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 4932416G01Rik; CAD13; cadherin 13; cadherin 13, H-cadherin (heart); cadherin-13; CD39 antigen-like 3; CD39L3; CD39-like 3; CDH13; CDHH; Cdht; ecto-apyrase 3; ecto-ATPase 3; ecto-ATPDase 3; HB6; H-cadherin; H-cadherin (heart); heart cadherin; heart-cadherin; NTPDase 3; NTPDase-3; P105; T cad; T Cadherin; Tcad; T-cad; T-cadherin; Truncated cadherin; truncated-cadherin
Assay Range 40 to 10,000 pg/mL
Assay Sensitivity 40 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 1012
Gene Symbol CDH13
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target H-cadherin
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.