Learn More
ACROBiosystems Human Cadherin-17 / CDH17 Protein, Mouse IgG1 Fc Tag
Supplier:  ACROBiosystems CA7H52A9100UG
Encompass_Preferred
Human Cadherin-17 Protein, Mouse IgG1 Fc Tag (CA7-H52A9) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Met 787 (Accession # Q12864-1).
Catalog No. 50-233-1603