Learn More
ACROBiosystems ACROBiosystems Human Cadherin-6 / KCAD / CDH6 Protein, His Tag
Supplier:  ACROBiosystems CA6H52291MG
Encompass_Preferred
Recombinant Protein;1MG;Human Cadherin-6, His Tag (CA6-H5229) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Ala 615 (Accession # AAH00019).;This protein carries a polyhistidine tag at the C-terminus.The Human Cadherin-6 will be further processed into mature form (Ser 54-Ala 615). The protein has a calculated MW of 67.2 kDa (pro-form) and 63.4 kDa (mature-form). The protein migrates as 86-96 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6536
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.