Learn More
R&D Systems™ Human Calbindin D DuoSet ELISA Kit
Description
Specifications
Specifications
| Assay Range | 31.2 to 2,000pg/mL |
| Conjugate | Biotin |
| Immunoassay Kit Format | 96-well Strip Plate |
| Product Type | Solid Phase Sandwich ELISA |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 795 (Human); 12309 (Mouse); 24249 (Rat) |
| Kit Contents | Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) |
| Quantity | 1 Kit |
| Sample Volume | 100 μL |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.