Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Calreticulin ELISA Kit

Catalog No. EH63RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH63RB 96 Tests
EH63RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH63RB Supplier Invitrogen™ Supplier No. EH63RB
Add to Cart
Add to Cart

ELISA

Human Calreticulin quantitates human Calreticulin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Calreticulin.

Calr or Calreticulin is a calcium-binding chaperone that promotes folding, oligomeric assembly and quality control in the endoplasmic reticulum (ER) via the calreticulin/calnexin cycle. Calr, a lectin, interacts transiently with almost all of the monoglucosylated glycoproteins that are synthesized in the ER. Calr also interacts with with the DNA-binding domain of NR3C1 and mediates its nuclear export. It may also be involved in maternal gene expression regulation and oocyte maturation via the regulation of calcium homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Autoantigen RO; CABP3; CALBP; Calcium-binding protein 3; CALR; calrectulin; calregulin; Calreticulin; cC1qR; CRP55; CRT; CRTC; Endoplasmic reticulum resident protein 60; epididymis secretory sperm binding protein Li 99 n; ERp60; FLJ26680; grp60; HACBP; HEL-S-99 n; I79
Assay Range 1.3 to 200 ng/mL
Assay Sensitivity 1.3 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 811
Gene Symbol CALR
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Calreticulin
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.