Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CALU Full-length ORF (NP_001210.1, 1 a.a. - 315 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89937877 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-937-877 25 ug
89-937-876 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-937-877 Supplier Abnova™ Supplier No. H00000813P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The product of this gene is a calcium-binding protein localized in the endoplasmic reticulum (ER) and it is involved in such ER functions as protein folding and sorting. This protein belongs to a family of multiple EF-hand proteins (CERC) that include reticulocalbin, ERC-55, and Cab45 and the product of this gene. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MDLRQFLMCLSLCTAFALSKPTEKKDRVHHEPQLSDKVHNDAQSFDYDHDAFLGAEEAKTFDQLTPEESKERLGKIVSKIDGDKDGFVTVDELKDWIKFAQKRWIYEDVERQWKGHDLNEDGLVSWEEYKNATYGYVLDDPDPDDGFNYKQMMVRDERRFKMADKDGDLIATKEEFTAFLHPEEYDYMKDIVVQETMEDIDKNADGFIDLEEYIGDMYSHDGNTDEPEWVKTEREQFVEFRDKNRDGKMDKEETKDWILPSDYDHAEAEARHLVYESDQNKDGKLTKEEIVDKYDLFVGSQATDFGEALVRHDEF

Specifications

Accession Number NP_001210.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 813
Molecular Weight (g/mol) 63.5kDa
Name CALU (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MDLRQFLMCLSLCTAFALSKPTEKKDRVHHEPQLSDKVHNDAQSFDYDHDAFLGAEEAKTFDQLTPEESKERLGKIVSKIDGDKDGFVTVDELKDWIKFAQKRWIYEDVERQWKGHDLNEDGLVSWEEYKNATYGYVLDDPDPDDGFNYKQMMVRDERRFKMADKDGDLIATKEEFTAFLHPEEYDYMKDIVVQETMEDIDKNADGFIDLEEYIGDMYSHDGNTDEPEWVKTEREQFVEFRDKNRDGKMDKEETKDWILPSDYDHAEAEARHLVYESDQNKDGKLTKEEIVDKYDLFVGSQATDFGEALVRHDEF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ90608
Common Name CALU
Gene Symbol CALU
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.