Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CAMK1D (NM_153498) Full-length Recombinant Protein with GST-tag expressed in Sf9 cells

Catalog No. 89937887 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-937-887 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-937-887 Supplier Abnova™ Supplier No. P3844

Used for Func, SDS-PAGE

This gene encodes a member of the Ca2+/calmodulin-dependent protein kinase 1 subfamily of serine/threonine kinases. The encoded protein may be involved in the regulation of granulocyte function through the chemokine signal transduction pathway. Alternatively spliced transcript variants encoding different isoforms of this gene have been described. [provided by RefSeq]

Specifications

Accession Number NM_153498
For Use With (Application) Functional Study, SDS-PAGE
Formulation In 50mM Tris-HCl, 150mM NaCl, pH 7.5 (0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol)
Gene ID (Entrez) 57118
Molecular Weight (g/mol) 68kDa
Name CAMK1D (Human) Recombinant Protein
Preparation Method Insect cell (Sf9) expression system
Quality Control Testing SDS-PAGE Stained with Coomassie Blue
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CKLiK/CaM-K1/CaMKID
Common Name CAMK1D
Gene Symbol CAMK1D
Biological Activity The specific activity was determined to be 138nmol/min/mg as per Activity Assay Protocol.
Species Baculovirus
Recombinant Recombinant
Protein Tag GST
Expression System Insect cell (Sf9) expression system
Purity or Quality Grade >90% by densitometry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.