Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CAPN11 Partial ORF (NP_008989, 557 a.a. - 651 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89937957 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-937-957 25 μg
89-937-956 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-937-957 Supplier Abnova™ Supplier No. H00011131Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Calpains constitute a family of intracellular calcium-dependent cysteine proteases. There are eight members in this superfamily. They consist of a variable 80 kDa subunit and an invariant 30 kDa subunit. This calpain protein appears to have protease activity and calcium-binding ability. A similar mouse protein may play a functional role in spermatogenesis and in the regulation of calcium-dependent signal transduction events during meiosis. [provided by RefSeq]

Sequence: LLNRMAIKFKSFKTKGFGLDACRCMINLMDKDGSGKLGLLEFKILWKKLKKWMDIFRECDQDHSGTLNSYEMRLVIEKAGIKLNNKVMQVLVARY

Specifications

Accession Number NP_008989
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 11131
Molecular Weight (g/mol) 36.19kDa
Name CAPN11 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen LLNRMAIKFKSFKTKGFGLDACRCMINLMDKDGSGKLGLLEFKILWKKLKKWMDIFRECDQDHSGTLNSYEMRLVIEKAGIKLNNKVMQVLVARY
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias calpain11
Common Name CAPN11
Gene Symbol CAPN11
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.