Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Carbohydrate Sulfotransferase 3/CHST3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7450
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7450 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7450 Supplier R&D Systems Supplier No. MAB7450
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Carbohydrate Sulfotransferase 3/CHST3 Monoclonal specifically detects Carbohydrate Sulfotransferase 3/CHST3 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Carbohydrate Sulfotransferase 3/CHST3
Applications Immunohistochemistry
Classification Monoclonal
Clone 799011
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q7LGC8
Gene Alias C6ST1, C6ST-1, C6ST1HSD, C6STEC 2.8.2.17, carbohydrate (chondroitin 6) sulfotransferase 3, carbohydrate sulfotransferase 3, Chondroitin 6-O-sulfotransferase 1, Chondroitin 6-sulfotransferase, CHST3, EC 2.8.2, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 0, GST-0
Gene Symbols CHST3
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Carbohydrate Sulfotransferase 3/CHST3 Glu39-Thr479 Accession # Q7LGC8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9469
Test Specificity Detects human Carbohydrate Sulfotransferase 3/CHST3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human CHST1 or recombinant mouse CHST3 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.