Learn More
ACROBiosystems Recombinant Protein;Human Carbonic Anhydrase IX / CA9 (38-414) Protein;HEK293;1MG;CA9,CAIX,CA9,CA-IX,G250,MN,Carbonic anhydrase 9,P54/58N,pMW1
Supplier:  ACROBiosystems CA9H52261MG
Encompass_Preferred
Recombinant Protein;Human Carbonic Anhydrase IX / CA9 (38-414) Protein;1MG;Human CA9, His Tag (CA9-H5226) is expressed from human 293 cells (HEK293). It contains AA Gln 38 - Asp 414 (Accession # NP_001207).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 41.2 kDa. The protein migrates as 53-54 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. NC1555793
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.