Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Carbonic Anhydrase IX Quantikine ELISA Kit
SDP

Catalog No. DCA900 Shop All Novus Biologicals Products
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
DCA900 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DCA900 Supplier R&D Systems™ Supplier No. DCA900

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Carbonic Anhydrase IX Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Carbonic Anhydrase IX Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Carbonic Anhydrase IX/CA9 in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul), urine (100 ul).

Specifications

Assay Range 15.60 - 1000pg/mL
Assay Sensitivity 4.39pg/mL
Conjugate HRP
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine
Specificity Natural and Recombinant Human CA9
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 768
Sample Volume Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Citrate Plasma (100uL),Urine (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CA9, CAIX, CA-IX, Carbonate dehydratase IX, carbonic anhydrase 9, carbonic anhydrase IX, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MN, P54/58 N, PMW1, RCC, RCC-associated antigen G250, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.