Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Carbonic Anhydrase XIII/CA13 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7060956
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2194SP 25 μg
AF2194 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2194SP Supplier R&D Systems Supplier No. AF2194SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Carbonic Anhydrase XIII/CA13 Polyclonal specifically detects Carbonic Anhydrase XIII/CA13 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Carbonic Anhydrase XIII/CA13
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8N1Q1
Gene Alias CA13, carbonic anhydrase 13, carbonic anhydrase XIIICarbonate dehydratase XIII, CAXIII, CA-XIII, EC 4.2.1.1, FLJ37995, MGC59868
Gene Symbols CA13
Host Species Goat
Immunogen E. coli-derived recombinant human Carbonic Anhydrase XIII/CA13 Ser2-His262 Accession # Q8N1Q1
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 377677
Test Specificity Detects human Carbonic Anhydrase XIII/CA13 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant human (rh) CA1, rhCA2, and rhCA3 is observed and 5% cross-reactivity with rhCA4, rhCA8, rhCA9, rhCA10, rhCA12, and rhCA14 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.