Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Carboxylesterase 2/CES2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5657
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5657 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5657 Supplier R&D Systems Supplier No. MAB5657
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Carboxylesterase 2/CES2 Monoclonal specifically detects Carboxylesterase 2/CES2 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunocytochemistry.

Specifications

Antigen Carboxylesterase 2/CES2
Applications Western Blot, Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone 720711
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O00748
Gene Alias carboxylesterase 2 (intestine, liver), carboxylesterase 2EC 3.1.1.1, CE-2, CE-2PCE-2, CES2, CES2A1, EC 3.1.1, EC 3.1.1.84, hCE-2, iCE, intestinal carboxylesterase; liver carboxylesterase-2, PCE-2
Gene Symbols CES2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Carboxylesterase 2/CES2 Gln27-Leu559 (predicted) Accession # O00748
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8824
Test Specificity Detects human Carboxylesterase 2/CES2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse Carboxylesterase 2 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.