Learn More
Abnova™ Human CARD9 Full-length ORF (AAH08877.1, 1 a.a. - 492 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
The protein encoded by this gene is a member of the CARD protein family, which is defined by the presence of a characteristic caspase-associated recruitment domain (CARD). CARD is a protein interaction domain known to participate in activation or suppression of CARD containing members of the caspase family, and thus plays an important regulatory role in cell apoptosis. This protein was identified by its selective association with the CARD domain of BCL10, a postive regulator of apoptosis and NF-kappaB activation, and is thought to function as a molecular scaffold for the assembly of a BCL10 signaling complex that activates NF-kappaB. Several alternatively spliced transcript variants have been observed, but their full-length nature is not clearly defined. [provided by RefSeq]
Specifications
Specifications
| Accession Number | AAH08877.1 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 64170 |
| Molecular Weight (g/mol) | 83.1kDa |
| Name | CARD9 (Human) Recombinant Protein (P01) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 25 μg |
| Immunogen | MSDYENDDECWSVLEGFRVTLTSVIDPSRITPYLRQCKVLNPDDEEQVLSDPNLVIRKRKVGVLLDILQRTGHKGYVAFLESLELYYPQLYKKVTGKEPARVFSMIIDASGESGLTQLLMTEVMKLQKKVQDLTALLSSKDDFIKELRVKDSLLRKHQERVQRLKEECEAGSRELKRCKEENYDLAMRLAHQSEEKGAALMRNRDLQLEIDQLKHSLMKAEDDCKVERKHTLKLRHAMEQRPSQELLWELQQEKALLQARVQELEASVQEGKLDRSSPYIQVLEEDWRQALRDHQEQANTIFSLRKDLRQGEARRLRCMEEKEMFELQCLALRKDSKMYKDRIEAILLQMEEVAIERDQAIATREELHAQHARGLQEKDALRKQVRELGEKADELQLQVFQCEAQLLAVEGRLRRQQLETLVLSSDLEDGSPRRSQELSLPQDLEDTQLSDKGCLAGGGSPKQPFAALHQEQVLRNPHDAGPAGLPGIGAVC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.