Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Cardiac Myoglobin DuoSet ELISA
SDP

Catalog No. DY12505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY12505 1 Kit (for 5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY12505 Supplier R&D Systems™ Supplier No. DY12505
Add to Cart
Add to Cart
This item is not returnable. View return policy

DuoSet ELISA

Specifications

Assay Range 0.31 - 20 ng/mL
For Use With (Application) ELISA
Gene ID (Entrez) 4151
Quantity 1 Kit (for 5 Plates)
Storage Requirements Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Synonym MB, MGC13548, myoglobin, PVALB
Target Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.