Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Cardiotrophin-1 Recombinant Protein, PeproTech®

Catalog No. 30032250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30032250UG 250 μg
30032100UG 100 μg
300-32-10UG 10 μg
300-32-1MG 1 mg
300-32-2UG 2 μg
30032500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 30032250UG Supplier Gibco™ Supplier No. 30032250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human CT-1 is a 21.1 kDa protein consisting of 200 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Cardiotrophin-1 (CT-1) is a member of the IL-6 family of cytokines, which signal through gp130 receptor complexes. gp130 complexes with several different receptor subunits to transmit signals from Cardiotrophin-1, IL-6, LIF, OSM, CNTF and IL-11. Cardiotrophin-1 binds to and activates the leukemia inhibitory factor (LIF) receptor/gp130 receptor complex and has been shown to induce hypertrophy in cardiac myocytes in vitro. Cardiotrophin-1, a secreted protein expressed at high levels in myocardium during cardiogenesis, has been shown to promote proliferation and survival of embryonic cardiomyocytes, suggesting a role for Cardiotrophin-1 in the activation of gp130 during cardiac development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16619
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 1489
Molecular Weight (g/mol) 21.1 kDa
Name Human Cardiotrophin-1
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias cardiophin 1; Cardiotrophin; cardiotrophin 1; Cardiotrophin1; cardiotrophin-1; CT 1; CT1; CT-1; CT-1 CT1; CTF 1; Ctf1; IL-6 cytokine family
Common Name Cardiotrophin 1
Gene Symbol CTF1
Biological Activity The ED50 as determined by the dose-dependent proliferation of TF-1 cells was ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10^6 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence SRREGSLEDP QTDSSVSLLP HLEAKIRQTH SLAHLLTKYA EQLLQEYVQL QGDPFGLPSF SPPRLPVAGL SAPAPSHAGL PVHERLRLDA AALAALPPLL DAVCRRQAEL NPRAPRLLRR LEDAARQARA LGAAVEALLA ALGAANRGPR AEPPAATASA ASATGVFPAK VLGLRVCGLY REWLSRTEGD LGQLLPGGSA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.