Learn More
ACROBiosystems Human Catenin beta-1 200ug
Supplier:  ACROBiosystems CA1H81E3200UG
Encompass_Preferred
Biotinylated Human Catenin beta-1 HisAvitag (CA1-H81E3) is expressed from E. coli cells. It contains AA Asn 138 - Glu 686 (Accession # P35222-1).
Catalog No. 50-253-5448
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.