Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CBLB Full-length ORF (AAH32851.1, 1 a.a. - 982 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89938165 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10μg
25μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-938-165 10μg
89-938-166 25μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-938-165 Supplier Abnova™ Supplier No. H00000868P01S

Used for AP, Array, ELISA, WB-Re

Sequence: MANSMNGRNPGGRGGNPRKGRILGIIDAIQDAVGPPKQAAADRRTVEKTWKLMDKVVRLCQNPKLQLKNSPPYILDILPDTYQHLRLILSKYDDNQKLAQLSENEYFKIYIDSLMKKSKRAIRLFKEGKERMYEEQSQDRRNLTKLSLIFSHMLAEIKAIFPNGQFQGDNFRITKADAAEFWRKFFGDKTIVPWKVFRQCLHEVHQISSGLEAMALKSTIDLTCNDYISVFEFDIFTRLFQPWGSILRNWNFLAVTHPGYMAFLTYDEVKARLQKYSTKPGSYIFRLSCTRLGQWAIGYVTGDGNILQTIPHNKPLFQALIDGSREGFYLYPDGRSYNPDLTGLCEPTPHDHIKVTQEQYELYCEMGSTFQLCKICAENDKDVKIEPCGHLMCTSCLTAWQESDGQGCPFCRCEIKGTEPIIVDPFDPRDEGSRCCSIIDPFGMPMLDLDDDDDREESLMMNRLANVRKCTDRQNSPVTSPGSSPLAQRRKPQPDPLQIPHLSLPPVPPRLDLIQKGIVRSPCGSPTGSPKSSPCMVRKQDKPLPAPPPPLRDPPPPPPERPPPIPPDNRLSRHIHHVESVPSKDPPMPLEAWCPRDVFGTNQLVGCRLLGEGSPKPGITASSNVNGRHSRVGSDPVLMRKHRRHDLPLEGAKVFSNGHLGSEEYDVPPRLSPPPPVTTLLPSIKCTGPLANSLSEKTRDPVEEDDDEYKIPSSHPVSLNSQPSHCHNVKPPVRSCDNGHCMLNGTHGPSSEKKSNIPDLSIYLKGDVFDSASDPVPLPPARPPTRDNPKHGSSLNRTPSDYDLLIPPLGEDAFDALPPSLPPPPPPARHSLIEHSKPPGSSSRPSSGQDLFLLPSDPFVDLASGQVPLPPARRLPGENVKTNRTSQDYDQLPSCSDGSQAPARPPKPRPRRTAPEIHHRKPHGPEAALENVDAKIAKLMGEGYAFEEVKRALEIAQNNVEVARSILREFAFPPPVSPRLNL

Specifications

Accession Number AAH32851.1
For Use With (Application) Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 868
Molecular Weight (g/mol) 135.8kDa
Name Human CBLB Full-length ORF (AAH32851.1, 1 a.a. - 982 a.a.) Recombinant Protein with GST-tag at N-terminal
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10μg
Storage Requirements Store at −80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686J10223, DKFZp779A0729, DKFZp779F1443, FLJ36865, FLJ41152, Nbla00127, RNF56
Common Name CBLB
Gene Symbol CBLB
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST Tag
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.