Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CCBL1 Full-length ORF (AAH13069, 1 a.a. - 112 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89938219 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-938-219 25 ug
89-938-218 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-938-219 Supplier Abnova™ Supplier No. H00000883P02L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a cytosolic enzyme that is responsible for the metabolism of cysteine conjugates of certain halogenated alkenes and alkanes. This metabolism can form reactive metabolites leading to nephrotoxicity and neurotoxicity. Increased levels of this enzyme have been linked to schizophrenia. Multiple transcript variants that encode different isoforms have been identified for this gene. [provided by RefSeq]

Sequence: MRHHTRPIFLFLVETGFSHVGQAGLELLTSGDPPVSASQSAGIIGVSHRTWPCSSDFDAALGAGLELTLLPRLVVGTSLSPPEPRPAGPRGRVETCPSSSPGLGGHPCLHLL

Specifications

Accession Number AAH13069
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 883
Molecular Weight (g/mol) 38.06kDa
Name CCBL1 (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MRHHTRPIFLFLVETGFSHVGQAGLELLTSGDPPVSASQSAGIIGVSHRTWPCSSDFDAALGAGLELTLLPRLVVGTSLSPPEPRPAGPRGRVETCPSSSPGLGGHPCLHLL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ95217/GTK/KAT1/KATI/MGC29624
Common Name CCBL1
Gene Symbol CCBL1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.