Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CCDC58 (aa 4-71) Control Fragment Recombinant Protein

Catalog No. RP100171
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP100171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP100171 Supplier Invitrogen™ Supplier No. RP100171
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61587 (PA5-61587. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CCDC58 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q4VC31
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 131076
Name Human CCDC58 (aa 4-71) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A930007B11Rik; AI413631; CCDC58; coiled-coil domain containing 58; coiled-coil domain-containing protein 58; RGD1564582
Common Name CCDC58
Gene Symbol CCDC58
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PSGGVNCEEFAEFQELLKVMRTIDDRIVHELNTTVPTASFAGKIDASQTCKQLYESLMAAHASRDRVI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.