Learn More
ACROBiosystems Human CCL1 20ug
Supplier:  ACROBiosystems CC1H525320UG
Encompass_Preferred
Human CCL1 / I-309 Protein Mouse IgG2a Fc Tag (CC1-H5253) is expressed from human 293 cells (HEK293). It contains AA Lys 24 - Lys 96 (Accession # P22362-1).
Catalog No. 50-253-5407
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.