Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CCL15 (MIP-5) Recombinant Protein, PeproTech®

Catalog No. 30043500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30043500UG 500 μg
30043100UG 100 μg
300-43-1MG 1 mg
30043250UG 250 μg
300-43-25UG 25 μg
300-43-50UG 50 μg
300-43-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 missing translation for 'options'
This item is not returnable. View return policy
Catalog No. 30043500UG missing translation for 'mfr' Gibco™ missing translation for 'supplierNo' 30043500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human MIP-5 is a 10.1 kDa protein containing 92 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene is located in a cluster of similar genes in the same region of chromosome 17. These genes encode CC cytokines, which are secreted proteins characterized by two adjacent cysteines. The product of this gene is chemotactic for T cells and monocytes, and acts through C-C chemokine receptor type 1 (CCR1). The proprotein is further processed into numerous smaller functional peptides. Naturally-occurring readthrough transcripts occur from this gene into the downstream gene, CCL14 (chemokine (C-C motif) ligand 14).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16663
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 6359
Molecular Weight (g/mol) 10.1 kDa
Name Human CCL15 (MIP-5)
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C-C motif chemokine 15; C-C motif chemokine ligand 15; CCL15; CCL15(22-92); CCL15(25-92); CCL15(29-92); chemokine (C-C motif) ligand 15; chemokine CC-2; HCC-2; HCC-2 Lkn-1; HMRP-2 B; leukotactin 1; Leukotactin-1; LKN1; LKN-1; Macrophage inflammatory protein 5; MIP-1 delta; MIP-1 D; MIP5; MIP-5; MIP-5 MIP-1 d; Mrp-2 b; NCC3; NCC-3; NCC-3 MIP-1 D; NCC3MRP-2 B; new CC chemokine 3; SCYA15; SCYA15member 15; SCYL3; small inducible cytokine subfamily A (Cys-Cys), member 15; small-inducible cytokine A15; SY15
Common Name MIP-1 delta
Gene Symbol CCL15
Biological Activity Determined by its ability to chemoattract human T lymphocytes using a concentration range of 1.0-10.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence QFTNDAETEL MMSKLPLENP VVLNSFHFAA DCCTSYISQS IPCSLMKSYF ETSSECSKPG VIFLTKKGRQ VCAKPSGPGV QDCMKKLKPY SI
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.