Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CCL18 (MIP-4) Recombinant Protein, PeproTech®

Catalog No. 300342UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
300-34-2UG 2 μg
30034100UG 100 μg
300-34-10UG 10 μg
300-34-1MG 1 mg
30034250UG 250 μg
30034500UG 500 μg
300-34-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 300-34-2UG Supplier Gibco™ Supplier No. 300342UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human MIP-4 is a 7.8 kDa protein containing 68 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Macrophage Inflammatory Protein-4 (MIP-4), also called CCL18, is a chemokine expressed in the lymph nodes, lungs, placenta and bone marrow. Although the MIP-4s receptor remains undetermined, MIP-4 is known to act as a chemoattractant for activated and non-activated T cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P55774
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 6362
Molecular Weight (g/mol) 7.8 kDa
Name Human CCL18 (MIP-4)
Quantity 2 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Alternative macrophage activation-associated CC chemokine 1; AMAC1; AMAC-1; CC chemokine ligand 18; CC chemokine PARC; C-C motif chemokine 18; C-C motif chemokine ligand 18; CCL18; CCL18(1-68); CCL18(3-69); CCL18(4-69); chemokine (C-C motif) ligand 18 (pulmonary and activation-regulated); chemokine (C-C), dendritic; CKb7; CUL9; CUL-9; cullin 9; cullin-9; DCCK1; DC-CK1; dendritic cell chemokine 1; H7AP1; H-MIP-4; KIAA0708; Macrophage inflammatory protein 4; MIP4; MIP-4; p53-associated parkin-like cytoplasmic protein; PARC; parkin-like cytoplasmic p53 binding protein; pulmonary and activation-regulated chemokine; SCYA18; small inducible cytokine A18; small inducible cytokine subfamily A (Cys-Cys), member 18, pulmonary and activation-regulated; small-inducible cytokine A18; UbcH7-associated protein 1
Common Name PARC
Gene Symbol CCL18
Conjugate Unconjugated
Recombinant Recombinant
Sequence QVGTNKELCC LVYTSWQIPQ KFIVDYSETS PQCPKPGVIL LTKRGRQICA DPNKKWVQKY ISDLKLNA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.