Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CCL19 (MIP-3 beta) Recombinant Protein, PeproTech®

Catalog No. 30029B500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
20 μg
250 μg
5 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30029B500UG 500 μg
30029B100UG 100 μg
30029B20UG 20 μg
30029B250UG 250 μg
300-29B-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 30029B500UG Supplier Gibco™ Supplier No. 30029B500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

300-29B-1MG will be provided as 2 x 500 μg (300-29B-500UG). Recombinant Human MIP-3beta is an 8.8 kDa protein containing 77 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CCL19, also known as MIP-3 beta (Macrophage Inflammatory Protein 3 beta), is a member of the CC- subfamily of chemokines. It is most closely related to CCL21, with which it shares 32% amino acid sequence homology. CCL19 and CCL21 are expressed mainly by stromal cells in the T cell-rich zones of lymph nodes. They are critical mediators of the homeostatic trafficking of naive T cells and activated dendritic cells into the secondary lymphoid organs. CCL19 and CCL21 also play a role in T cell priming and activation, as well as the recruitment of lymphocytes to inflamed tissue. Both proteins signal via the G protein-coupled receptor, CCR7, which is expressed on T cells and mature dendritic cells. Although CCL19 and CCL21 both exhibit the same affinity for CCR7, only the binding of CCL19 results in the desensitization and internalization of the receptor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99731
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 6363
Molecular Weight (g/mol) 8.8 kDa
Name Human CCL19 (MIP-3 beta)
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias beta chemokine exodus-3; beta-chemokine exodus-3; CC chemokine ligand 19; C-C motif chemokine; C-C motif chemokine 19; C-C motif chemokine ligand 19; Ccl19; chemokine (C-C motif) ligand 19; chemokine CCL19; CK beta-11; CKb11; EBI1 ligand chemokine; EBI-1 ligand chemokine; EBI1-ligand chemokine; EBV-induced molecule 1 ligand chemokine; ELC; Epstein-Barr virus-induced molecule 1 ligand chemokine; exodus-3; macrophage inflammatory protein 3 beta; macrophage inflammatory protein 3-beta; MIP3B; MIP-3 b; MIP-3-beta; M-MIP-3-beta; SCYA19; small inducible cytokine A19; small inducible cytokine subfamily A (Cys-Cys), member 19; small-inducible cytokine A19
Common Name CCL19
Gene Symbol Ccl19
Biological Activity Determined by its ability to chemoattract human T cells using a concentration of 10.0-50.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence GTNDAEDCCL SVTQKPIPGY IVRNFHYLLI KDGCRVPAVV FTTLRGRQLC APPDQPWVER IIQRLQRTSA KMKRRSS
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.