Learn More
Invitrogen™ Human CCL2 (MCP-1) Recombinant Protein
Description
CCL2 (human) Recombinant Protein, RP-8648, from Invitrogen.
Specifications
Specifications
| Accession Number | P13500 |
| Concentration | 0.1mg/mL |
| For Use With (Application) | Control |
| Formulation | 0.1%TFA with no preservative |
| Gene ID (Entrez) | 6347 |
| Molecular Weight (g/mol) | 8.6 |
| Name | Human CCL2 (MCP-1) |
| Quantity | 20 μg |
| Source | E. coli |
| Storage Requirements | -20°C, Avoid Freeze/Thaw Cycles |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.